missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methoxycoumarin, 98+%, ACROS Organics™

Chemical Identifiers

CAS 531-59-9
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00006876
Chemical Name or Material 7-Methoxycoumarin
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC205120500 SDS Thermo Fisher Scientific
205120500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

531-59-9
100.0
C10H8O3
MFCD00006876
≥98.0%
98.0
98+%
176.17
7-Methoxycoumarin

Specifications

Powder
176.17
117° to 121°C
98.0
Glass bottle
MFCD00006876
98+%
7-Methoxycoumarin
Beige to salmon-pinkish
50g
18, 27
100.0
C10H8O3
531-59-9
Authentic
≥98.0%