Please login to your online account to display your discounted pricing

7-Methoxycoumarin, 98+%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  205120500

 View more versions of this product

Catalog No. AC205120500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Color Beige to salmon-pinkish
Assay ≥98.0%
Formula Weight 176.17
Quantity 50g
Melting Point 117° to 121°C
Beilstein 18, 27
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 7-Methoxycoumarin
Molecular Formula C10H8O3
MDL Number MFCD00006876
CAS 531-59-9
Assay Percent Range 98+%
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 7-Methoxycoumarin
CAS 531-59-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00006876