Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxycoumarin, 98+%

$40.82 - $465.59

Chemical Identifiers

Chemical Name or Material 7-Methoxycoumarin
CAS 531-59-9
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00006876
Beilstein 141728
Synonym Ayapanin; Herniarin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1544106

  SDS
alfa aesar™
A1544106
5g Each for $40.82

AAA1544114

  SDS
alfa aesar™
A1544114
25g Each for $134.72

AAA1544122

  SDS
alfa aesar™
A1544122
100g Each for $465.59
Description & Specifications

Specifications

Formula Weight 176.17
TSCA TSCA
Quantity 5g
Assay ≥98%
Melting Point 117° to 120°C
Chemical Name or Material 7-Methoxycoumarin
RTECS Number DJ3100380
Beilstein 141728
Synonym Ayapanin; Herniarin
CAS 531-59-9
MDL Number MFCD00006876
Molecular Formula C10H8O3

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.