missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxybenzo[b]furan-2-carboxylic acid, 97%

$287.22 - $287.22

Chemical Identifiers

CAS 4790-79-8
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00060516
Chemical Name or Material 7-Methoxybenzo[b]furan-2-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2118506 SDS Alfa Aesar™
B2118506
5g Each for $287.22
AAB2118503 SDS Alfa Aesar™
B2118503
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4790-79-8
192.17
7-Methoxybenzo[b]furan-2-carboxylic acid
C10H8O4
MFCD00060516

Specifications

217° to 220°C
4790-79-8
192.17
7-Methoxybenzo[b]furan-2-carboxylic acid
5g
C10H8O4
MFCD00060516
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning