missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxy-4-methylcoumarin, 99%

Manufacturer:  Alfa Aesar™ A1178503

 View more versions of this product

Catalog No. AAA1178503


 

Description

Description

Applications
A coumarin derivative and fluorescent label.

Solubility
Soluble in Methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

7-Methoxy-4-methylcoumarin
1g
99%
MFCD00009773
Methyl 4-methylumbelliferyl ether
CC1=CC(=O)OC2=C1C=CC(=C2)OC
190.198
190.2
159° to 162°C
2555-28-4
C11H10O3
156900
UDFPKNSWSYBIHO-UHFFFAOYSA-N
7-methoxy-4-methylchromen-2-one
390807

Chemical Identifiers

2555-28-4
190.198
UDFPKNSWSYBIHO-UHFFFAOYSA-N
390807
C11H10O3
MFCD00009773
Methyl 4-methylumbelliferyl ether
SDS Documents