missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methoxy-4-methylcoumarin, 98%, ACROS Organics™

Chemical Identifiers

CAS 2555-28-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H10O3
Formula Weight 190.2
MDL Number MFCD00009773
Chemical Name or Material 7-Methoxy-4-methylcoumarin
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC334110010 SDS Thermo Fisher Scientific
334110010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC334112500 SDS Thermo Fisher Scientific
334112500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2555-28-4
100.0
C11H10O3
MFCD00009773
≥97.5%
97.5
98%
190.2
7-Methoxy-4-methylcoumarin

Specifications

190.2
1g
157.0° to 161.0°C
C11H10O3
2555-28-4
MFCD00009773
Authentic
≥97.5%
Powder, Crystals, Crystalline Powder and/or Chunks
White to yellow
100.0
98%
97.5
Glass bottle
7-Methoxy-4-methylcoumarin