Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxy-2-tetralone, 98%

$49.57 - $49.57

Chemical Identifiers

Chemical Name or Material 7-Methoxy-2-tetralone
CAS 4133-34-0
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001730
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2243203

 
alfa aesar™
B2243203
1g Each for $49.57
Description & Specifications

Specifications

Formula Weight 176.22
Chemical Name or Material 7-Methoxy-2-tetralone
Assay 98%
Molecular Formula C11H12O2
MDL Number MFCD00001730
Boiling Point 124° to 126°C (1.5mmHg)
CAS 4133-34-0
Quantity 1g
Flash Point >110°C (230°F)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C