missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methoxy-2-tetralone, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC294970250

 View more versions of this product

Catalog No. AC294970250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7-Methoxy-2-tetralone
>110°C
Glass bottle
1.13
Yellow to Orange
4133-34-0
100.0
C11H12O2
2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 3,4-dihydro-7-methoxynaphthalen-2 1h-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one
COC1=CC2=C(CCC(=O)C2)C=C1
176.22
176.22
95%
1.1300g/mL
Authentic
1.5590 to 1.563
124°C to 126°C (1.5mmHg)
25g
94.0
94% min. (GC)
MFCD00001730
XEAPZXNZOJGVCZ-UHFFFAOYSA-N
7-methoxy-3,4-dihydro-1H-naphthalen-2-one
77785
Liquid After Melting

Chemical Identifiers

4133-34-0
176.22
XEAPZXNZOJGVCZ-UHFFFAOYSA-N
77785
COC1=CC2=C(CCC(=O)C2)C=C1
C11H12O2
MFCD00001730
2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 3,4-dihydro-7-methoxynaphthalen-2 1h-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one
7-methoxy-3,4-dihydro-1H-naphthalen-2-one
Safety and Handling

Safety and Handling

EINECSNumber : 223-954-1

SDS Documents