Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxy-1-tetralone, 99%

$24.72 - $72.77

Chemical Identifiers

Chemical Name or Material 7-Methoxy-1-tetralone
CAS 6836-19-7
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001696
Beilstein 2208389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1513003

  SDS
alfa aesar™
L1513003
1g Each for $24.72

AAL1513006

  SDS
alfa aesar™
L1513006
5g Each for $72.77
Description & Specifications

Specifications

Formula Weight 176.22
CAS 6836-19-7
MDL Number MFCD00001696
Molecular Formula C11H12O2
Beilstein 2208389
Quantity 1g
Melting Point 59° to 63°C
Chemical Name or Material 7-Methoxy-1-tetralone
Assay 99%