Please login to your online account to display your discounted pricing

7-Methoxy-1-tetralone, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC172430050

 View more versions of this product

Catalog No. AC172430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 176.21
Quantity 5g
Color White to beige
Melting Point 61° to 64°C
Beilstein 09, II, 889
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 7-Methoxy-1-tetralone
Assay Percent Range 97%
Molecular Formula C11H12O2
CAS 6836-19-7
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Infrared Spectrum Authentic
MDL Number MFCD00001696
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Packaging Glass bottle

Specifications

Chemical Name or Material 7-Methoxy-1-tetralone
CAS 6836-19-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H12O2
Formula Weight 176.21
MDL Number MFCD00001696
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents