missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methoxy-1-naphthaleneacetic Acid, TCI America™

$160.42 - $481.27

Chemical Identifiers

CAS 6836-22-2
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.236
MDL Number MFCD12756806
InChI Key GJPYHKXILUFWKV-UHFFFAOYSA-N
Synonym (7-Methoxy-1-naphthyl)acetic Acid
PubChem CID 13570105
IUPAC Name 2-(7-methoxynaphthalen-1-yl)acetic acid
SMILES COC1=CC2=C(C=CC=C2CC(=O)O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28885G View Documents TCI America
M28885G
5g Each for $481.27
 
M28881G View Documents TCI America
M28881G
1g Each for $160.42
 
Specifications

Chemical Identifiers

6836-22-2
216.236
GJPYHKXILUFWKV-UHFFFAOYSA-N
13570105
COC1=CC2=C(C=CC=C2CC(=O)O)C=C1
C13H12O3
MFCD12756806
(7-Methoxy-1-naphthyl)acetic Acid
2-(7-methoxynaphthalen-1-yl)acetic acid

Specifications

White-Yellow
5g
C13H12O3
(7-Methoxy-1-naphthyl)acetic Acid
COC1=CC2=C(C=CC=C2CC(=O)O)C=C1
216.236
216.24
≥98.0% (GC,T)
157°C
6836-22-2
MFCD12756806
GJPYHKXILUFWKV-UHFFFAOYSA-N
2-(7-methoxynaphthalen-1-yl)acetic acid
13570105
Crystal-Powder at 20°C
7-Methoxy-1-naphthaleneacetic Acid
Safety and Handling

Safety and Handling

TSCA : No