missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxy-1-indanone, 95%

$82.96 - $263.70

Chemical Identifiers

CAS 34985-41-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD06659666
InChI Key CZXBVBATQPHSSL-UHFFFAOYSA-N
Synonym 7-methoxy-1-indanone, 7-methoxy-2,3-dihydro-1h-inden-1-one, 7-methoxyindan-1-one, 2,3-dihydro-7-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-7-methoxy, 7-methoxy-indan-1-one, 2,3-dihydro-7-methoxy-1h-inden-1-one, 1h-inden-1-one,2,3-dihydro-7-methoxy, 7-methoxyindanone, acmc-209unh
PubChem CID 288143
IUPAC Name 7-methoxy-2,3-dihydroinden-1-one
SMILES COC1=CC=CC2=C1C(=O)CC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66249MD View Documents Alfa Aesar™
H66249MD
250mg Each for $82.96
 
AAH6624903 View Documents Alfa Aesar™
H6624903
1g Each for $263.70
 
Specifications

Chemical Identifiers

34985-41-6
162.188
CZXBVBATQPHSSL-UHFFFAOYSA-N
288143
COC1=CC=CC2=C1C(=O)CC2
C10H10O2
MFCD06659666
7-methoxy-1-indanone, 7-methoxy-2,3-dihydro-1h-inden-1-one, 7-methoxyindan-1-one, 2,3-dihydro-7-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-7-methoxy, 7-methoxy-indan-1-one, 2,3-dihydro-7-methoxy-1h-inden-1-one, 1h-inden-1-one,2,3-dihydro-7-methoxy, 7-methoxyindanone, acmc-209unh
7-methoxy-2,3-dihydroinden-1-one

Specifications

99°C to 102°C
34985-41-6
MFCD06659666
7-methoxy-1-indanone, 7-methoxy-2,3-dihydro-1h-inden-1-one, 7-methoxyindan-1-one, 2,3-dihydro-7-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-7-methoxy, 7-methoxy-indan-1-one, 2,3-dihydro-7-methoxy-1h-inden-1-one, 1h-inden-1-one,2,3-dihydro-7-methoxy, 7-methoxyindanone, acmc-209unh
COC1=CC=CC2=C1C(=O)CC2
162.188
162.19
95%
250mg
C10H10O2
907449
CZXBVBATQPHSSL-UHFFFAOYSA-N
7-methoxy-2,3-dihydroinden-1-one
288143
Solid
7-Methoxy-1-indanone
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P301+P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning