missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Methoxy-1-indanone, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H66249MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
7-Methoxy-1-indanone | |
99°C to 102°C | |
MFCD06659666 | |
907449 | |
CZXBVBATQPHSSL-UHFFFAOYSA-N | |
7-methoxy-2,3-dihydroinden-1-one | |
288143 | |
95% |
34985-41-6 | |
C10H10O2 | |
250 mg | |
7-methoxy-1-indanone, 7-methoxy-2,3-dihydro-1h-inden-1-one, 7-methoxyindan-1-one, 2,3-dihydro-7-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-7-methoxy, 7-methoxy-indan-1-one, 2,3-dihydro-7-methoxy-1h-inden-1-one, 1h-inden-1-one,2,3-dihydro-7-methoxy, 7-methoxyindanone, acmc-209unh | |
COC1=CC=CC2=C1C(=O)CC2 | |
162.188 | |
162.19 | |
Solid |
Chemical Identifiers
34985-41-6 | |
162.188 | |
CZXBVBATQPHSSL-UHFFFAOYSA-N | |
7-methoxy-2,3-dihydroinden-1-one |
C10H10O2 | |
MFCD06659666 | |
288143 | |
COC1=CC=CC2=C1C(=O)CC2 |
Safety and Handling
GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
H302-H317-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only