Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxy-1,4-benzothiazin-3(4H)-one, 97%

$61.64 - $189.85

Chemical Identifiers

Chemical Name or Material 7-Methoxy-1,4-benzothiazin-3(4H)-one
CAS 22726-30-3
Molecular Formula C9H9NO2S
Formula Weight 195.24
MDL Number MFCD02660686
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2783703

  SDS
alfa aesar™
H2783703
1g Each for $61.64

AAH2783706

  SDS
alfa aesar™
H2783706
5g Each for $189.85
Description & Specifications

Specifications

Formula Weight 195.24
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD02660686
Molecular Formula C9H9NO2S
Quantity 1g
CAS 22726-30-3
Melting Point 182° to 184°C
Chemical Name or Material 7-Methoxy-1,4-benzothiazin-3(4H)-one
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.