missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-MAC 95.0+%, TCI America™

Manufacturer:  Tci America M21295G

 View more versions of this product

Catalog No. M21295G



Specifications

Specifications

7-MAC
120°C
56610-72-1
MFCD08704705
QGXKMJVEULWQSB-VWNXMTODSA-N
benzhydryl (6R,7S)-7-amino-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
12882026
Crystal-Powder at 20°C
Yellow
5g
C24H24N6O4S2
Diphenylmethyl (6R,7S)-7-Amino-7-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Diphenylmethyl 7beta-Amino-7alpha-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-3-cephem-4-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(N)OC)SC2)C(=O)OC(C4=CC=CC=C4)C5=CC=CC=C5
524.614
524.61
≥95.0% (HPLC)

Chemical Identifiers

56610-72-1
524.614
QGXKMJVEULWQSB-VWNXMTODSA-N
12882026
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(N)OC)SC2)C(=O)OC(C4=CC=CC=C4)C5=CC=CC=C5
C24H24N6O4S2
MFCD08704705
Diphenylmethyl (6R,7S)-7-Amino-7-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Diphenylmethyl 7beta-Amino-7alpha-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-3-cephem-4-carboxylate
benzhydryl (6R,7S)-7-amino-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions