missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxyisoflavone, TCI America™

$250.56 - $750.16

Chemical Identifiers

CAS 13057-72-2
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00017701
InChI Key WMKOZARWBMFKAS-UHFFFAOYSA-N
Synonym 7-Hydroxy-3-phenylchromone
PubChem CID 5376891
ChEBI CHEBI:12256
IUPAC Name 7-hydroxy-3-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15795G View Documents TCI America
H15795G
5g Each for $750.16
 
H15791G View Documents TCI America
H15791G
1g Each for $250.56
 
Specifications

Chemical Identifiers

13057-72-2
238.242
WMKOZARWBMFKAS-UHFFFAOYSA-N
5376891
7-hydroxy-3-phenylchromen-4-one
C15H10O3
MFCD00017701
7-Hydroxy-3-phenylchromone
CHEBI:12256
C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O

Specifications

White-Yellow
5g
C15H10O3
7-Hydroxy-3-phenylchromone
C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O
238.242
CHEBI:12256
Crystal-Powder at 20°C
7-Hydroxyisoflavone
216°C
13057-72-2
MFCD00017701
WMKOZARWBMFKAS-UHFFFAOYSA-N
7-hydroxy-3-phenylchromen-4-one
5376891
238.24
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : DJ3100255

TSCA : No