missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxyflavanone 98.0+%, TCI America™

Supplier:  TCI America H10061G

 View more versions of this product

Catalog No. H10061G



Description

Description

Specifications

Specifications

7-Hydroxyflavanone
190°C
MFCD00017487
7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3
240.258
CHEBI:34483
≥98.0% (HPLC,T)
6515-36-2
C15H12O3
1 g
SWAJPHCXKPCPQZ-UHFFFAOYSA-N
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
1890
240.26
Crystalline Powder

Chemical Identifiers

6515-36-2
240.258
SWAJPHCXKPCPQZ-UHFFFAOYSA-N
1890
7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
C15H12O3
MFCD00017487
7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo
CHEBI:34483
C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions