missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxycoumarin, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 121110250

 View more versions of this product

Catalog No. AC121110250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

7-Hydroxycoumarin
0.5% max. (105°C, 3 hrs)
0.2% max.
228°C to 234°C
99%
98.5
99%
MFCD00006878
15, 10032
Solubility in water: 1g/100mL (100 c). Other solubilities: freely soluble in acetic acid and dilute alkalies,freely soluble in alcohol and chloroform,sparingly soluble in ether
C1=CC(=CC2=C1C=CC(=O)O2)O
162.14
CHEBI:27510
Powder and Agglomerate
Authentic
Plastic bottle
Beige-Brown to White
25g
93-35-6
100.0
C9H6O3
18, V,1, 386
7-hydroxy-2h-chromen-2-one, 7-hydroxycoumarin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangin, hydrangine, skimmetin, skimmetine, umbelliferon, umbelliferone
ORHBXUUXSCNDEV-UHFFFAOYSA-N
7-hydroxychromen-2-one
5281426
162.14
99%

Chemical Identifiers

93-35-6
162.14
ORHBXUUXSCNDEV-UHFFFAOYSA-N
5281426
7-hydroxychromen-2-one
C9H6O3
MFCD00006878
7-hydroxy-2h-chromen-2-one, 7-hydroxycoumarin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangin, hydrangine, skimmetin, skimmetine, umbelliferon, umbelliferone
CHEBI:27510
C1=CC(=CC2=C1C=CC(=O)O2)O
Safety and Handling

Safety and Handling

EINECSNumber : 202-240-3

RTECSNumber : GN6820000

SDS Documents