missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxycoumarin-4-acetic Acid 98.0+%, TCI America™

$82.30 - $279.80

Chemical Identifiers

CAS 6950-82-9
Molecular Formula C11H8O5
Molecular Weight (g/mol) 220.18
MDL Number MFCD00037563
InChI Key BNHPMQBVNXMPDU-UHFFFAOYSA-N
Synonym 7-hydroxycoumarin-4-acetic acid, 2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid, umbelliferone-4-acetic acid, 7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid, chembl12253, 2-7-hydroxy-2-oxochromen-4-yl acetic acid, 7-hydroxy-4-coumarinylacetic acid, 2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo, 7-hydroxycoumarin-4-yl acetic acid, 7-hydroxy-2-oxochromen-4-yl acetic acid
PubChem CID 5338490
IUPAC Name 2-(7-hydroxy-2-oxochromen-4-yl)acetic acid
SMILES C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15011G View Documents Tci America
H15011G
1g Each for $82.30
 
H15015G View Documents Tci America
H15015G
5g Each for $279.80
 
Specifications

Chemical Identifiers

6950-82-9
220.18
BNHPMQBVNXMPDU-UHFFFAOYSA-N
5338490
C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O
C11H8O5
MFCD00037563
7-hydroxycoumarin-4-acetic acid, 2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid, umbelliferone-4-acetic acid, 7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid, chembl12253, 2-7-hydroxy-2-oxochromen-4-yl acetic acid, 7-hydroxy-4-coumarinylacetic acid, 2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo, 7-hydroxycoumarin-4-yl acetic acid, 7-hydroxy-2-oxochromen-4-yl acetic acid
2-(7-hydroxy-2-oxochromen-4-yl)acetic acid

Specifications

White-Yellow
1g
C11H8O5
7-hydroxycoumarin-4-acetic acid, 2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid, umbelliferone-4-acetic acid, 7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid, chembl12253, 2-7-hydroxy-2-oxochromen-4-yl acetic acid, 7-hydroxy-4-coumarinylacetic acid, 2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo, 7-hydroxycoumarin-4-yl acetic acid, 7-hydroxy-2-oxochromen-4-yl acetic acid
C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O
220.18
220.18
≥98.0% (HPLC,T)
190°C
6950-82-9
MFCD00037563
BNHPMQBVNXMPDU-UHFFFAOYSA-N
2-(7-hydroxy-2-oxochromen-4-yl)acetic acid
5338490
Crystal-Powder at 20°C
7-Hydroxycoumarin-4-acetic Acid
Safety and Handling

Safety and Handling

TSCA : No