Please login to your online account to display your discounted pricing

7-Hydroxy-PIPAT maleate, Tocris

D3 agonist (D3 > D2)

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-PIPAT maleate
Assay Percent Range >98%
Molecular Formula C16H22INO·C4H4O4
Formula Weight 487.33
Synonym (RS)-trans-7-Hydroxy-2-[N-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

071910

 
r & d systems
RDS071910
10mg Each for $221.01

071950

 
r & d systems
RDS071950
50mg Each for $904.70
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 487.33
Chemical Name or Material 7-Hydroxy-PIPAT maleate
CAS 200722-46-9
Synonym (RS)-trans-7-Hydroxy-2-[N-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin maleate
Molecular Formula C16H22INO·C4H4O4
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C