Please login to your online account to display your discounted pricing

7-Hydroxy-PIPAT maleate, Tocris


D3 agonist (D3 > D2)

Quantity 50mg

Manufacturer: r & d systems  RDS071950

 View more versions of this product

Catalog No. 071950

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 487.33
Chemical Name or Material 7-Hydroxy-PIPAT maleate
Molecular Formula C16H22INO·C4H4O4
Assay Percent Range >98%
CAS 200722-46-9
Synonym (RS)-trans-7-Hydroxy-2-[N-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin maleate
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 7-Hydroxy-PIPAT maleate
Assay Percent Range >98%
Molecular Formula C16H22INO·C4H4O4
Formula Weight 487.33
Synonym (RS)-trans-7-Hydroxy-2-[N-propyl-N-(3'-iodo-2'-propenyl)amino]tetralin maleate
Accessories

Desiccate at +4°C