Please login to your online account to display your discounted pricing

7-Hydroxy-DPAT hydrobromide, Tocris


Dopamine agonist (D3 ≥ D2 < > D4)

Quantity 10mg

Manufacturer: r & d systems  RDS070610

 View more versions of this product

Catalog No. 070610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 328.28
Chemical Name or Material 7-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
CAS 76135-30-3
Synonym (±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 7-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
Formula Weight 328.28
Synonym (±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
Accessories

Desiccate at +4°C