Please login to your online account to display your discounted pricing

7-Hydroxy-DPAT hydrobromide, Tocris

Dopamine agonist (D3 ≥ D2 < > D4)

$89.94 - $336.06

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
Formula Weight 328.28
Synonym (±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

070610

 
r & d systems
RDS070610
10mg Each for $89.94

070650

 
r & d systems
RDS070650
50mg Each for $336.06
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 328.28
Chemical Name or Material 7-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
CAS 76135-30-3
Synonym (±)-7-Hydroxy-2-dipropylaminotetralin hydrobromide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C