Learn More
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 277822500
Specifications
7-Hydroxy-6-methoxy-3, 4-dihydroisoquinoline | |
187°C to 191°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
MFCD00143517 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in chloroform,ethanol,methanol and hot,ethyl acetate | |
COC1=C(C=C2C=NCCC2=C1)O | |
177.2 | |
177.2 |
Glass bottle | |
250mg | |
4602-73-7 | |
100.0 | |
C10H11NO2 | |
3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, ksc496q9t, pubchem12735 | |
UQBWYWCIBNWMPI-UHFFFAOYSA-N | |
6-methoxy-3,4-dihydroisoquinolin-7-ol | |
363043 | |
98% |
Chemical Identifiers
4602-73-7 | |
177.2 | |
UQBWYWCIBNWMPI-UHFFFAOYSA-N | |
363043 | |
COC1=C(C=C2C=NCCC2=C1)O |
C10H11NO2 | |
MFCD00143517 | |
3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, ksc496q9t, pubchem12735 | |
6-methoxy-3,4-dihydroisoquinolin-7-ol |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning