missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC277820010

 View more versions of this product

Catalog No. AC277820010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

7-Hydroxy-6-methoxy-3, 4-dihydroisoquinoline
187°C to 191°C
98%
97.5
97.5% min. (GC)
MFCD00143517
Solubility in water: soluble in hot water. Other solubilities: soluble in chloroform,ethanol,methanol and hot,ethyl acetate
COC1=C(C=C2C=NCCC2=C1)O
177.2
177.2
Glass bottle
1g
4602-73-7
100.0
C10H11NO2
3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, ksc496q9t, pubchem12735
UQBWYWCIBNWMPI-UHFFFAOYSA-N
6-methoxy-3,4-dihydroisoquinolin-7-ol
363043
98%

Chemical Identifiers

4602-73-7
177.2
UQBWYWCIBNWMPI-UHFFFAOYSA-N
363043
COC1=C(C=C2C=NCCC2=C1)O
C10H11NO2
MFCD00143517
3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, ksc496q9t, pubchem12735
6-methoxy-3,4-dihydroisoquinolin-7-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents