Please login to your online account to display your discounted pricing

7-Hydroxy-4-(trifluoromethyl)-2H-chromen-2-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
CAS 575-03-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H5F3O3
Formula Weight 230.14
MDL Number MFCD00037578
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RJF00002EA

 
fisher scientific company
RJF00002EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.14
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
Molecular Formula C10H5F3O3
MDL Number MFCD00037578
CAS 575-03-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.