Please login to your online account to display your discounted pricing

7-hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid, Maybridge

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H12O5
Formula Weight 248.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10150DE

 
fisher scientific company
BTB10150DE
Amber glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB10150DA

  SDS
fisher scientific company
BTB10150DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.23
Quantity 5g
Assay 97%
CAS 105212-09-7
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C13H12O5
Chemical Name or Material 7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning