Please login to your online account to display your discounted pricing

7-hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB10150DA

 View more versions of this product

Catalog No. BTB10150DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 248.23
Quantity 1g
Assay 97%
Chemical Name or Material 7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
Assay Percent Range 97%
Molecular Formula C13H12O5
CAS 105212-09-7
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 7-Hydroxy-4-oxo-8-propyl-4H-chromene-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H12O5
Formula Weight 248.23
Accessories

GHS Signal Word: Warning

SDS & Safety Documents