missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Hydroxy-4'-nitroisoflavone, 98%

Manufacturer:  Alfa Aesar™ H6416722

 View more versions of this product

Catalog No. AAH6416722



Description

Description

Applications
7-Hydroxy-4′-nitroisoflavone is used in the synthesis of iso flavones, Synthesis of daidzin, soflavone amide derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

7-Hydroxy-4'-nitroisoflavone
290°C to 292°C
15485-80-0
MFCD01546432
7-hydroxy-4'-nitroisoflavone, 7-hydroxy-3-4-nitrophenyl-4h-chromen-4-one, 7-hydroxy-3-4-nitrophenyl chromen-4-one, 7-hydroxy-3-4-nitrophenyl-4h-1-benzopyran-4-one, 7-hydroxy-4-nitroisoflavone, 7-hydroxy-4/'-nitroisoflavone, 7-hydroxy-4'-nitro-isoflavone 3c
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)[N+](=O)[O-]
283.239
283.24
98%
Yellow
100g
C15H9NO5
385006
CEESSYSSRMOMDE-UHFFFAOYSA-N
7-hydroxy-3-(4-nitrophenyl)chromen-4-one
7454026
Powder

Chemical Identifiers

15485-80-0
283.239
CEESSYSSRMOMDE-UHFFFAOYSA-N
7454026
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)[N+](=O)[O-]
C15H9NO5
MFCD01546432
7-hydroxy-4'-nitroisoflavone, 7-hydroxy-3-4-nitrophenyl-4h-chromen-4-one, 7-hydroxy-3-4-nitrophenyl chromen-4-one, 7-hydroxy-3-4-nitrophenyl-4h-1-benzopyran-4-one, 7-hydroxy-4-nitroisoflavone, 7-hydroxy-4/'-nitroisoflavone, 7-hydroxy-4'-nitro-isoflavone 3c
7-hydroxy-3-(4-nitrophenyl)chromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents