missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Hydroxy-4-methyl-8-nitrocoumarin, 98%

Manufacturer:  Alfa Aesar™ L0789603

Catalog No. AAL0789603



Specifications

Specifications

7-Hydroxy-4-methyl-8-nitrocoumarin
1g
C10H7NO5
225171
BGUBUSIGKOWDPO-UHFFFAOYSA-N
7-hydroxy-4-methyl-8-nitrochromen-2-one
5376327
98%
∽255°C (decomposition)
19037-69-5
MFCD00051824
2h-1-benzopyran-2-one, 7-hydroxy-4-methyl-8-nitro, 2h-1-benzopyran-2-one,7-hydroxy-4-methyl-8-nitro, 7-hydroxy-4-methyl-8-nitro-2h-chromen-2-one, 7-hydroxy-4-methyl-8-nitro-2h-chromen-2-one #, 7-hydroxy-4-methyl-8-nitrocoumarin, 7-hydroxy-8-nitro-4-methyl-3-chromene-2-one, 7-hydroxy-8-nitro-4-methylcoumarin, acmc-20alwz, pubchem8690
CC1=CC(=O)OC2=C1C=CC(=C2[N+](=O)[O-])O
221.168
221.17

Chemical Identifiers

19037-69-5
221.168
BGUBUSIGKOWDPO-UHFFFAOYSA-N
5376327
CC1=CC(=O)OC2=C1C=CC(=C2[N+](=O)[O-])O
C10H7NO5
MFCD00051824
2h-1-benzopyran-2-one, 7-hydroxy-4-methyl-8-nitro, 2h-1-benzopyran-2-one,7-hydroxy-4-methyl-8-nitro, 7-hydroxy-4-methyl-8-nitro-2h-chromen-2-one, 7-hydroxy-4-methyl-8-nitro-2h-chromen-2-one #, 7-hydroxy-4-methyl-8-nitrocoumarin, 7-hydroxy-8-nitro-4-methyl-3-chromene-2-one, 7-hydroxy-8-nitro-4-methylcoumarin, acmc-20alwz, pubchem8690
7-hydroxy-4-methyl-8-nitrochromen-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents