missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Hydroxy-4-(methoxymethyl)coumarin, 98%

$121.80 - $453.28

Chemical Identifiers

CAS 157101-77-4
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00052607
InChI Key PRVOZGKAZHXKMM-UHFFFAOYSA-N
Synonym 2h-1-benzopyran-2-one, 7-hydroxy-4-methoxymethyl, 7-hydroxy-4-methoxymethyl chromen-2-one, 7-hydroxy-4-methoxymethyl coumarin, 7-hydroxy-4-methoxymethyl-2h-chromen-2-one, 7-hydroxy-4-methoxymethyl-chromen-2-one, 7-hydroxy-4-methoxymethylcoumarin, acmc-20alx1
PubChem CID 5393168
IUPAC Name 7-hydroxy-4-(methoxymethyl)chromen-2-one
SMILES COCC1=CC(=O)OC2=C1C=CC(=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1856203 View Documents Alfa Aesar™
A1856203
1g Each for $121.80
 
AAA1856206 View Documents Alfa Aesar™
A1856206
5g Each for $453.28
 
Specifications

Chemical Identifiers

157101-77-4
206.197
PRVOZGKAZHXKMM-UHFFFAOYSA-N
5393168
COCC1=CC(=O)OC2=C1C=CC(=C2)O
C11H10O4
MFCD00052607
2h-1-benzopyran-2-one, 7-hydroxy-4-methoxymethyl, 7-hydroxy-4-methoxymethyl chromen-2-one, 7-hydroxy-4-methoxymethyl coumarin, 7-hydroxy-4-methoxymethyl-2h-chromen-2-one, 7-hydroxy-4-methoxymethyl-chromen-2-one, 7-hydroxy-4-methoxymethylcoumarin, acmc-20alx1
7-hydroxy-4-(methoxymethyl)chromen-2-one

Specifications

126°C to 128°C
157101-77-4
MFCD00052607
PRVOZGKAZHXKMM-UHFFFAOYSA-N
7-hydroxy-4-(methoxymethyl)chromen-2-one
5393168
98%
1g
C11H10O4
2h-1-benzopyran-2-one, 7-hydroxy-4-methoxymethyl, 7-hydroxy-4-methoxymethyl chromen-2-one, 7-hydroxy-4-methoxymethyl coumarin, 7-hydroxy-4-methoxymethyl-2h-chromen-2-one, 7-hydroxy-4-methoxymethyl-chromen-2-one, 7-hydroxy-4-methoxymethylcoumarin, acmc-20alx1
COCC1=CC(=O)OC2=C1C=CC(=C2)O
206.197
206.2
7-Hydroxy-4-(methoxymethyl)coumarin
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures