Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 97%

$44.45 - $44.45

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone
CAS 22246-18-0
Molecular Formula C9H9NO2
Formula Weight 163.17
MDL Number MFCD06410891
Synonym 7-Hydroxy-3,4-dihydrocarbostyril
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2748503

  SDS
alfa aesar™
H2748503
1g Each for $44.45
Description & Specifications

Specifications

Formula Weight 163.17
Chemical Name or Material 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone
Assay 97%
Molecular Formula C9H9NO2
MDL Number MFCD06410891
CAS 22246-18-0
Synonym 7-Hydroxy-3,4-dihydrocarbostyril
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 230° to 234°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.