missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide

Selective agonist for D3 dopamine

$186.43 - $777.36

Chemical Identifiers

CAS 74938-11-7
Molecular Formula C16H26BrNO
Molecular Weight (g/mol) 328.294
MDL Number MFCD00153809
InChI Key ODNDMTWHRYECKX-UHFFFAOYSA-N
Synonym +/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, 7-oh-dpat hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, r +-7-hydroxy-dpat hydrobromide, y-7-hydroxy-dpat hydrobromide
PubChem CID 11957566
IUPAC Name 7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
SMILES CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61065MA View Documents Alfa Aesar™
J61065MA
10mg Each for $186.43
 
AAJ61065MF View Documents Alfa Aesar™
J61065MF
50mg Each for $777.36
 
Specifications

Chemical Identifiers

74938-11-7
328.294
ODNDMTWHRYECKX-UHFFFAOYSA-N
11957566
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
C16H26BrNO
MFCD00153809
+/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, 7-oh-dpat hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, r +-7-hydroxy-dpat hydrobromide, y-7-hydroxy-dpat hydrobromide
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide

Specifications

White
10mg
C16H26BrNO
+/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, 7-oh-dpat hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, r +-7-hydroxy-dpat hydrobromide, y-7-hydroxy-dpat hydrobromide
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
328.294
329.29
(+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide
168°C to 170°C
74938-11-7
MFCD00153809
ODNDMTWHRYECKX-UHFFFAOYSA-N
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
11957566
Powder
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures