Alfa Aesar™ (+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide
Selective agonist for D3 dopamine
Manufacturer:  Alfa Aesar™ J61065MA
 View more versions of this product
Catalog No. AAJ61065MA
Specifications
Specifications
(+/-)-7-Hydroxy-2-di-n-propylaminotetralin hydrobromide
168°C to 170°C
74938-11-7
MFCD00153809
ODNDMTWHRYECKX-UHFFFAOYSA-N
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
11957566
Powder
White
10mg
C16H26BrNO
7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-oh-dpat hydrobromide, r +-7-hydroxy-dpat hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, y-7-hydroxy-dpat hydrobromide
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
328.294
329.29
Chemical Identifiers
74938-11-7
328.294
ODNDMTWHRYECKX-UHFFFAOYSA-N
11957566
CCCN(CCC)C1CCC2=C(C1)C=C(C=C2)O.Br
C16H26BrNO
MFCD00153809
7-dipropylamino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide, 7-hydroxy-dpat hydrobromide, +/--7-oh-dpat, 7-oh-dpat hydrobromide, r +-7-hydroxy-dpat hydrobromide, +/--7-hydroxy-dpat hydrobromide, +/--7-hydroxy-2-di-n-propylamino tetralin hydrobromide, +/--7-hydroxy-2-dipropylaminotetralin hydrobromide, r +-7-hydroxy dipropylamino tetralin hydrobromide, y-7-hydroxy-dpat hydrobromide
7-(dipropylamino)-5,6,7,8-tetrahydronaphthalen-2-ol;hydrobromide
Documents