Please login to your online account to display your discounted pricing

7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one
CAS 50544-72-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H14O3
Formula Weight 206.24
MDL Number MFCD01314121
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH01364EA

 
fisher scientific company
RH01364EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RH01364DA

 
fisher scientific company
RH01364DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.24
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD01314121
CAS 50544-72-4
Molecular Formula C12H14O3