Please login to your online account to display your discounted pricing

7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH01364EA

 View more versions of this product

Catalog No. RH01364EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.24
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD01314121
CAS 50544-72-4
Molecular Formula C12H14O3

Specifications

Chemical Name or Material 7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one
CAS 50544-72-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H14O3
Formula Weight 206.24
MDL Number MFCD01314121