Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Hexadecyn-1-ol, 95%

$54.03 - $745.01

Chemical Identifiers

Chemical Name or Material 7-Hexadecyn-1-ol
CAS 822-21-9
Molecular Formula C16H30O
Linear Formula CH3(CH2)7C≡C(CH2)6OH
Formula Weight 238.41
MDL Number MFCD00041709
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2211303

 
alfa aesar™
B2211303
1g Each for $54.03

AAB2211306

 
alfa aesar™
B2211306
5g Each for $181.64

AAB2211314

 
alfa aesar™
B2211314
25g Each for $745.01
Description & Specifications

Specifications

Formula Weight 238.41
Chemical Name or Material 7-Hexadecyn-1-ol
Assay 95%
Molecular Formula C16H30O
Linear Formula CH3(CH2)7C≡C(CH2)6OH
MDL Number MFCD00041709
CAS 822-21-9
Quantity 1g

Applications
7-Hexadecyn-1-ol is used in the preparation of 15-hexadecynoic acid. It acts as a reagent for pharmaceutical research and in chemical synthesis.

Notes
Incompatible with strong oxidizing agents.