Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluoroisatin, 97%

$88.91 - $370.66

Chemical Identifiers

Chemical Name or Material 7-Fluoroisatin
CAS 317-20-4
Molecular Formula C8H4FNO2
Formula Weight 165.12
MDL Number MFCD01569508
Synonym 7-Fluoro-1H-indole-2,3-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2682706

  SDS
alfa aesar™
H2682706
5g Each for $88.91

AAH2682714

  SDS
alfa aesar™
H2682714
25g Each for $370.66
Description & Specifications

Specifications

Formula Weight 165.12
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 7-Fluoroisatin
Melting Point 193° to 195°C
Assay 97%
Molecular Formula C8H4FNO2
MDL Number MFCD01569508
CAS 317-20-4
Quantity 5g
Synonym 7-Fluoro-1H-indole-2,3-dione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

Solubility
Sparingly Soluble in water (0.022 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.