Learn More
7-Fluoroisatin, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2682706
Description
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.
Solubility
Sparingly Soluble in water (0.022 g/L)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
7-Fluoroisatin | |
193°C to 195°C | |
MFCD01569508 | |
7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion | |
HGBGVEOXPHGSOS-UHFFFAOYSA-N | |
7-fluoro-1H-indole-2,3-dione | |
586418 | |
97% |
317-20-4 | |
C8H4FNO2 | |
5 g | |
Sparingly Soluble in water (0.022g/L) | |
C1=CC2=C(C(=C1)F)NC(=O)C2=O | |
165.123 | |
165.12 |
Chemical Identifiers
317-20-4 | |
165.123 | |
HGBGVEOXPHGSOS-UHFFFAOYSA-N | |
586418 | |
C1=CC2=C(C(=C1)F)NC(=O)C2=O |
C8H4FNO2 | |
MFCD01569508 | |
7-fluoroisatin, 7-fluoroindoline-2,3-dione, 7-fluoro-2,3-indolinedione, 1h-indole-2,3-dione, 7-fluoro, 7-fluoroindole-1h-2,3-dione, 7-fluoroindole-2,3-dione, 7-fluoro-2,3-dihydro-1h-indole-2,3-dione, 7-fluoroisatine, 7-fluoro-1h-benzo d azoline-2,3-dione, 7-fluor-1h-indol-2,3-dion | |
7-fluoro-1H-indole-2,3-dione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only