Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluorobenzofurazan-4-sulfonamide, 98%

$661.47 - $1,952.76

Chemical Identifiers

Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonamide
CAS 91366-65-3
Molecular Formula C6H4FN3O3S
Formula Weight 217.17
MDL Number MFCD00036687
Synonym ABD-F; 4-Fluoro-7-sulfamoylbenzofurazan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB23586MB

  SDS
alfa aesar™
B23586MB
25mg Each for $661.47

AAB23586MC

  SDS
alfa aesar™
B23586MC
100mg Each for $1,952.76
Description & Specifications

Specifications

Formula Weight 217.17
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonamide
Assay 98%
CAS 91366-65-3
Quantity 25mg
Synonym ABD-F; 4-Fluoro-7-sulfamoylbenzofurazan
MDL Number MFCD00036687
Melting Point 143° to 147°C
Molecular Formula C6H4FN3O3S