missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%

$114.74 - $402.50

Chemical Identifiers

CAS 122138-83-4
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.153
MDL Number MFCD23379858
InChI Key PEOVRVSERLAIQM-UHFFFAOYSA-N
PubChem CID 14422956
IUPAC Name 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61065MD View Documents Alfa Aesar™
H61065MD
250mg Each for $114.74
 
AAH6106503 View Documents Alfa Aesar™
H6106503
1g Each for $402.50
 
Description

Description

Applications
This compounds are useful laboratory reagents and used frequently in organic synthesis.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Chemical Identifiers

122138-83-4
198.153
PEOVRVSERLAIQM-UHFFFAOYSA-N
7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
C8H7FN2O3
MFCD23379858
14422956
C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F

Specifications

250mg
C8H7FN2O3
PEOVRVSERLAIQM-UHFFFAOYSA-N
7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
14422956
97%
122138-83-4
MFCD23379858
C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F
198.153
198.15
7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning