Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Formula Weight 293.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6290003

 
alfa aesar™
H6290003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62900MD

 
alfa aesar™
H62900MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 293.1
Chemical Name or Material 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Physical Form Solid
Assay 96%
Quantity 1g