missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H62900MD

 View more versions of this product

Catalog No. AAH62900MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7-Fluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
2018286-71-8
C14H17BFNO4
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2F)OCC(=O)N3
293.101
293.1
250mg
96%
MIJNRXJMZCWQSY-UHFFFAOYSA-N
7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
75487208
Solid

Chemical Identifiers

2018286-71-8
293.101
75487208
C14H17BFNO4
MIJNRXJMZCWQSY-UHFFFAOYSA-N
Documents