Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Formula Weight 279.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6295503

 
alfa aesar™
H6295503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62955MD

 
alfa aesar™
H62955MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 279.11
Chemical Name or Material 7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Physical Form Solid
Assay 96%
Quantity 1g