missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Fluoro-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole, 95%, Alfa Aesar™

$211.30 - $674.66

Chemical Identifiers

CAS 177858-80-9
Molecular Formula C11H11FN2
Molecular Weight (g/mol) 190.221
MDL Number MFCD09264527
InChI Key OTGGOPSBNVEBDA-UHFFFAOYSA-N
Synonym 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole
PubChem CID 15077550
IUPAC Name 7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES C1CNCC2=C1C3=C(N2)C=C(C=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27780MC
View Documents
Thermo Fisher Scientific
H27780MC
100mg
Each for $211.30
 
AAH27780ME
View Documents
Thermo Fisher Scientific
H27780ME
500mg
Each for $674.66
 
Description

Description

Specifications

Chemical Identifiers

177858-80-9
190.221
OTGGOPSBNVEBDA-UHFFFAOYSA-N
15077550
C1CNCC2=C1C3=C(N2)C=C(C=C3)F
C11H11FN2
MFCD09264527
1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole
7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole

Specifications

216°C to 224°C
177858-80-9
MFCD09264527
OTGGOPSBNVEBDA-UHFFFAOYSA-N
7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
15077550
95%
100mg
C11H11FN2
1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole
C1CNCC2=C1C3=C(N2)C=C(C=C3)F
190.221
190.22
7-Fluoro-1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications