missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Ethylindole, 97%, ACROS Organics™

$172.21 - $172.21

Chemical Identifiers

CAS 22867-74-9
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00143514
InChI Key PIIZLMYXLGYWTN-UHFFFAOYSA-N
Synonym 7-ethylindole, 1h-indole, 7-ethyl, 7-ethylindole, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
PubChem CID 2724711
IUPAC Name 7-ethyl-1H-indole
SMILES CCC1=CC=CC2=C1NC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC271140250 SDS View Documents Acros Organics
271140250
25g Glass bottle Each for $172.21
 
Specifications

Chemical Identifiers

22867-74-9
145.205
PIIZLMYXLGYWTN-UHFFFAOYSA-N
2724711
CCC1=CC=CC2=C1NC=C2
C10H11N
MFCD00143514
7-ethylindole, 1h-indole, 7-ethyl, 7-ethylindole, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
7-ethyl-1H-indole

Specifications

1.05g/mL
Glass bottle
0.1% max. (K.F.)
25g
96.0
97%
MFCD00143514
PIIZLMYXLGYWTN-UHFFFAOYSA-N
7-ethyl-1H-indole
2724711
Liquid
7-Ethylindole
Authentic
1.6020 to 1.6040 (20°C, 589nm)
Undesignated
22867-74-9
100.0
C10H11N
7-ethylindole, 1h-indole, 7-ethyl, 7-ethylindole, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
CCC1=CC=CC2=C1NC=C2
145.205
145.2
≥96.0%