Please login to your online account to display your discounted pricing

7-Ethylindole, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  271140250

 View more versions of this product

Catalog No. AC271140250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 145.2
Physical Form Liquid
Quantity 25g
Color Clear yellow to red-brown
Density 1.05g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00143514
CAS 22867-74-9
Chemical Name or Material 7-Ethylindole
Molecular Formula C10H11N
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Water 0.1% max. (K.F.)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 7-Ethylindole
CAS 22867-74-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H11N
Formula Weight 145.2
MDL Number MFCD00143514