missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-(Ethylamino)-4,6-dimethylcoumarin, TCI America™

Manufacturer:  TCI America E1132200MG

 View more versions of this product

Catalog No. E1132200MG



Specifications

Specifications

7-(Ethylamino)-4,6-dimethylcoumarin
167°C
26078-25-1
MFCD00006860
QZXAEJGHNXJTSE-UHFFFAOYSA-N
7-(ethylamino)-4,6-dimethylchromen-2-one
96929
Crystal-Powder at 20°C
Undesignated
200mg
C13H15NO2
coumarin 2, 4,6-dimethyl-7-ethylamino coumarin, 7-ethylamino-4,6-dimethyl coumarin, 7-ethylamino-4,6-dimethyl-2h-chromen-2-one, 7-ethylamino-4,6-dimethyl-chromen-2-one, 4,6-dimethyl-7-ethylaminocoumarin, coumarin, 7-ethylamino-4,6-dimethyl, 2h-1-benzopyran-2-one, 7-ethylamino-4,6-dimethyl, coumarin 450, ccris 4958
CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C
217.268
217.27
≥98.0% (GC)

Chemical Identifiers

26078-25-1
217.268
QZXAEJGHNXJTSE-UHFFFAOYSA-N
96929
CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C
C13H15NO2
MFCD00006860
coumarin 2, 4,6-dimethyl-7-ethylamino coumarin, 7-ethylamino-4,6-dimethyl coumarin, 7-ethylamino-4,6-dimethyl-2h-chromen-2-one, 7-ethylamino-4,6-dimethyl-chromen-2-one, 4,6-dimethyl-7-ethylaminocoumarin, coumarin, 7-ethylamino-4,6-dimethyl, 2h-1-benzopyran-2-one, 7-ethylamino-4,6-dimethyl, coumarin 450, ccris 4958
7-(ethylamino)-4,6-dimethylchromen-2-one
Safety and Handling

Safety and Handling

TSCA : Yes

Documents