missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(Ethylamino)-4,6-dimethylcoumarin 98.0+%, TCI America™
Supplier: TCI America E11321G
Specifications
| 7-(Ethylamino)-4,6-dimethylcoumarin | |
| 167°C | |
| MFCD00006860 | |
| coumarin 2, 4,6-dimethyl-7-ethylamino coumarin, 7-ethylamino-4,6-dimethyl coumarin, 7-ethylamino-4,6-dimethyl-2h-chromen-2-one, 7-ethylamino-4,6-dimethyl-chromen-2-one, 4,6-dimethyl-7-ethylaminocoumarin, coumarin, 7-ethylamino-4,6-dimethyl, 2h-1-benzopyran-2-one, 7-ethylamino-4,6-dimethyl, coumarin 450, ccris 4958 | |
| CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C | |
| 217.268 | |
| 217.27 | |
| Crystalline Powder |
| 26078-25-1 | |
| C13H15NO2 | |
| 1 g | |
| QZXAEJGHNXJTSE-UHFFFAOYSA-N | |
| 7-(ethylamino)-4,6-dimethylchromen-2-one | |
| 96929 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 26078-25-1 | |
| 217.268 | |
| QZXAEJGHNXJTSE-UHFFFAOYSA-N | |
| 96929 | |
| CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C |
| C13H15NO2 | |
| MFCD00006860 | |
| coumarin 2, 4,6-dimethyl-7-ethylamino coumarin, 7-ethylamino-4,6-dimethyl coumarin, 7-ethylamino-4,6-dimethyl-2h-chromen-2-one, 7-ethylamino-4,6-dimethyl-chromen-2-one, 4,6-dimethyl-7-ethylaminocoumarin, coumarin, 7-ethylamino-4,6-dimethyl, 2h-1-benzopyran-2-one, 7-ethylamino-4,6-dimethyl, coumarin 450, ccris 4958 | |
| 7-(ethylamino)-4,6-dimethylchromen-2-one |
Safety and Handling
TSCA : Yes