missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Ethyl-3-indoleethanol, TCI America™

Manufacturer:  TCI America E08245G

 View more versions of this product

Catalog No. E08245G



Specifications

Specifications

7-Ethyl-3-indoleethanol
50°C
41340-36-7
MFCD01718805
7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol
CCC1=CC=CC2=C1NC=C2CCO
189.258
189.26
≥98.0% (GC)
White-Yellow
5g
C12H15NO
3077
UVSDNCAZVSQJQA-UHFFFAOYSA-N
2-(7-ethyl-1H-indol-3-yl)ethanol
198231
Crystal-Powder at 20°C

Chemical Identifiers

41340-36-7
189.258
UVSDNCAZVSQJQA-UHFFFAOYSA-N
198231
CCC1=CC=CC2=C1NC=C2CCO
C12H15NO
MFCD01718805
7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol
2-(7-ethyl-1H-indol-3-yl)ethanol
Safety and Handling

Safety and Handling

RTECSNumber : NL8512200

TSCA : No

Documents